missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KAT3B/p300 Antibody (RW105), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-616IR-0.1ml
This item is not returnable.
View return policy
Description
KAT3B/p300 Monoclonal antibody specifically detects KAT3B/p300 in Human, Mouse, Rat, Mustelid, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, In vitro Assay, Electron Microscopy, Flow (Intracellular), ChIP assay (ChIP), KnockDown
Specifications
| KAT3B/p300 | |
| Monoclonal | |
| DyLight 755 | |
| E1A binding protein p300, E1A-associated protein p300, E1A-binding protein, 300kD, EC 2.3.1, EC 2.3.1.48, histone acetyltransferase p300, KAT3B, P300, p300 HAT, RSTS2 | |
| Fusion protein containing residues 1572-2371 of human KAT3B/p300. [UniProt# Q0947] | |
| 0.1 mL | |
| Adaptive Immunity, Alzheimers Research, Amino Acids Drugs and other small molecules, Angiogenesis, Apoptosis, Asthma, Autophagy, Base Excision Repair, Beta Galactosidase Epitope Tags, Breast Cancer, Cancer, Cell Cycle and Replication, Cellular Markers, Chromatin Research, DNA Repair, HIF Target Genes, Hypoxia, Neuroscience, Stem Cell Markers, Transcription Factors and Regulators, Tumor Suppressors, Vision, Wnt Signaling Pathway | |
| 2033 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, In vitro Assay | |
| RW105 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Mustelid, Primate | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction