missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KAT3B/p300 Antibody (RW105), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-616H-0.1ml
This item is not returnable.
View return policy
Description
KAT3B/p300 Monoclonal antibody specifically detects KAT3B/p300 in Human, Mouse, Rat, Mustelid, Primate samples. It is validated for Western Blot, Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, In vitro Assay, Electron Microscopy, Flow (Intracellular), ChIP assay (ChIP), KnockDown
Specifications
| KAT3B/p300 | |
| Monoclonal | |
| HRP | |
| E1A binding protein p300, E1A-associated protein p300, E1A-binding protein, 300kD, EC 2.3.1, EC 2.3.1.48, histone acetyltransferase p300, KAT3B, P300, p300 HAT, RSTS2 | |
| Fusion protein containing residues 1572-2371 of human KAT3B/p300. [UniProt# Q0947] | |
| 0.1 mL | |
| Apoptosis, Autophagy, Breast Cancer, Cancer, Cell Cycle and Replication, Cellular Markers, Chromatin Research, DNA Repair, HIF Target Genes, Hypoxia, Stem Cell Markers, Transcription Factors and Regulators, Tumor Suppressors, Wnt Signaling Pathway | |
| 2033 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, In vitro Assay | |
| RW105 | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Mustelid, Primate | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction