missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KATNAL1 Antibody (OTI1A7), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72254H
This item is not returnable.
View return policy
Description
KATNAL1 Monoclonal specifically detects KATNAL1 in Human, Mouse, Rat, Primate, Monkey samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| KATNAL1 | |
| Monoclonal | |
| HRP | |
| PBS with No Preservative | |
| KATNAL1 | |
| Full length recombinant protein of human KATNAL1 (NP_115492) produced in HEK293T cells. | |
| 0.1 mL | |
| 84056.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry | |
| OTI1A7 | |
| Western Blot, Immunohistochemistry | |
| EC 3.6.4.3, katanin p60 ATPase-containing subunit A-like 1, katanin p60 subunit A-like 1MGC2599, p60 katanin-like 1 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat, Primate, Monkey | |
| IgG1 |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur