missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KATNB1 Monoclonal Antibody (OTI5E1), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503814
This item is not returnable.
View return policy
Description
Microtubules, polymers of alpha and beta tubulin subunits, form the mitotic spindle of a dividing cell and help to organize membranous organelles during interphase. Katanin is a heterodimer that consists of a 60 kDa ATPase (p60 subunit A 1) and an 80 kDa accessory protein (p80 subunit B 1). The p60 subunit acts to sever and disassemble microtubules, while the p80 subunit targets the enzyme to the centrosome. Katanin is a member of the AAA family of ATPases.Specifications
| KATNB1 | |
| Monoclonal | |
| 0.88 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9BVA0 | |
| KATNB1 | |
| Full length human recombit protein of human KATNB1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI5E1 | |
| Unconjugated | |
| KATNB1 | |
| KAT, LIS6 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 10300 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction