Learn More
Nordvlies Katrin Inclusive Gigant Toilet S Dispenser
Click to view available options
Color:
Black
White
Unit Size:
Each
Description
- Dispenser for big Gigant toilet rolls for high frequency usage
- The lock can be used with or without a key
- The whole paper roll can be used economically with integrated stub roll holder
- Easy paper tear-off to all sides and all edges
- No searching around inside the dispenser opening for the loose end of the paper roll
- Katrin Gigant S Dispensers are suitable for S sized Gigant rolls
- New flexible roll brake for better dispensing
- Main raw materials of major plastic parts and lock: ABS (Acrylonitrile butadiene styrene), PC (polycarbonate), POM (polyoxymethylene)
- Katrin Inclusive dispensers are resistant to high temperatures
- Comply with UL94 Fire Safety and Fire Protection regulations (EU)
Specifications
Specifications
| Product Type | Dispenser |
| Color | Black |
| Dimensions (L x W x H) | 245 x 239 x 151 mm |
| For Use With (Application) | Dispensing Hand Towels |
| Size | Small |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto