missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KAZALD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5119241
This item is not returnable.
View return policy
Description
KAZALD1 Polyclonal Antibody for Western Blot, IHC (P)
This gene encodes a secreted member of the insulin growth factor-binding protein (IGFBP) superfamily. It contains an N-terminal insulin growth factor-binding domain, a central Kazal-type serine protease inhibitor and follistatin-like domain, and a C-terminal immunoglobulin-like domain. Studies of the mouse ortholog suggest that this gene product may have a function in bone development and bone regeneration.
Specifications
| KAZALD1 | |
| Polyclonal | |
| Unconjugated | |
| KAZALD1 | |
| bone- and odontoblast-expressed gene 1; BONO1; FKSG28; FKSG40; FLJ24094; IGFBP-related protein 10; IGFBP-rP10; Kazal type serine peptidase inhibitor domain 1; KAZALD1; kazal-type serine protease inhibitor domain-containing protein 1; novel kazal-type serine protease inhibitor domain and immunoglobulin domain containing protein; UNQ2945/PRO21184 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 100624902, 617445, 81621 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q96I82 | |
| KAZALD1 | |
| Recombinant protein Kazal Type Serine Protease Inhibitor Domain Containing Protein 1. The antigen corresponds to amino acid range 31-304 of the target protein. | |
| 200 μL | |
| Primary | |
| Human, Bovine, Pig | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction