missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Kazrin Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA556926

Product Code. 15714831

  • € 578.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: RLQEEVHLLR QMKEMLAKDL EESQGGKSSE VLSATELRVQ LAQKEQELAR AKEALQAMKA DRKRLKGEKT DLVSQMQQLY ATLE Highest antigen sequence identity to the following orthologs: Mouse - 96%, Rat - 95%.

Component of the cornified envelope of keratinocytes. May be involved in the interplay between adherens junctions and desmosomes. The function in the nucleus is not known.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Kazrin
Polyclonal
Unconjugated
KAZN
2310007B04Rik; 9030409G11Rik; AI256607; AI666764; AW061099; HRIHFB2003; KAZ; KAZN; Kazrin; kazrin, periplakin interacting protein; KIAA1026
Rabbit
Antigen affinity chromatography
RUO
23254
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.3 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q674X7
KAZN
Recombinant protein corresponding to Human Kazrin. Recombinant protein control fragment (Product #RP-94541).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.