missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ KB SRC 4
GREENER_CHOICE

Product Code. p-4816566
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30001606 10 mg 10mg
30001624 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001606 Supplier Tocris Bioscience™ Supplier No. 4660/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective c-Src inhibitor

Potent and selective c-Src inhibitor (Ki = 44 nM); does not inhibit c-Abl at concentrations up to 125 μM. Selective between Src family members (Kd values are 86, 160, 240, 720, 3200, 4400, and >40,000 nM for c-Src, Lck, Fgr, Yes, Lyn, Hck and Fyn respectively). Significantly inhibits cell growth in 4T1 mammary carcinoma tumor cells.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1380088-03-8
Molecular Formula C32H23ClN8

Specifications

Quantity 10 mg
Formula Weight 555.03
Purity >98%
Molecular Weight (g/mol) 555.042
Chemical Name or Material KB SRC 4
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.