missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KBTBD12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5148415
This item is not returnable.
View return policy
Description
KBTBD12 Polyclonal Antibody for Western Blot
KBTBD12 is a protein coding gene.
Specifications
| KBTBD12 | |
| Polyclonal | |
| Unconjugated | |
| KBTBD12 | |
| 4833415F11Rik; 4933428M03Rik; KBTBD12; kelch domain containing 6; kelch domain-containing protein 6; kelch repeat and BTB (POZ) domain containing 12; kelch repeat and BTB domain containing 12; kelch repeat and BTB domain-containing protein 12; KLHDC6 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 166348 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 100 μg/mL | |
| PBS with 0.03% ProClin 300; pH 7 | |
| Q3ZCT8 | |
| KBTBD12 | |
| E. coli-derived Human KBTBD12 fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction