missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ KBTBD5 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17131534 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17131534 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17131534 Supplier Invitrogen™ Supplier No. PA5113673

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: ELVPTELNDI WRYNEEEKKW EGVLREIAYA AGATFLPVRL NVLCLTKM For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 93%; Dog: 93%; Guinea Pig: 86%; Horse: 93%; Human: 100%; Mouse: 93%; Pig: 93%; Rabbit: 100%; Rat: 79%.

The exact function of KBTBD5 remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KBTBD5
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Klhl40
Gene Accession No. Q2TBA0
Gene Alias 2310024D23Rik; KBTBD5; kelch like family member 40; kelch repeat and BTB (POZ) domain containing 5; kelch repeat and BTB domain-containing protein 5; kelch-like 40; kelch-like family member 40; kelch-like protein 40; kelch-like protein 40; LOW QUALITY PROTEIN: kelch-like protein 40; Klhl40; NEM8; nemaline myopathy type 8; sarcosynapsin; SRYP; SYRP
Gene Symbols Klhl40
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C terminal region of human KBTBD5.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 131377
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.