missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KBTBD7 Antibody (OTI1G3), DyLight 755, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72268IR
This item is not returnable.
View return policy
Description
KBTBD7 Monoclonal specifically detects KBTBD7 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| KBTBD7 | |
| Monoclonal | |
| DyLight 755 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| KBTBD7 | |
| Full length recombinant protein of human KBTBD7 (NP_115514) produced in HEK293T cells. | |
| 0.1 mL | |
| 84078.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| OTI1G3 | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| DKFZP434E2318, kelch repeat and BTB (POZ) domain containing 7, kelch repeat and BTB domain-containing protein 7 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction