missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KBTBD7 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113167
This item is not returnable.
View return policy
Description
KBTBD7 Polyclonal Antibody for Western Blot, IHC (P), ELISA
The protein encoded by this gene is a transcriptional activator, having been shown to increase the transcription of activator protein-1 and serum response element. The encoded protein can also form a complex with KBTBD6 and CUL3, which regulates the ubiquitylation and degradation of TIAM1, which is a regulator of RAC1.
Specifications
| KBTBD7 | |
| Polyclonal | |
| Unconjugated | |
| KBTBD7 | |
| KBTBD7; kelch repeat and BTB (POZ) domain containing 7; kelch repeat and BTB domain containing 7; kelch repeat and BTB domain-containing protein 7; kelch repeat and BTB domain-containing protein 7-like | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 84078 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 0.28 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8WVZ9 | |
| KBTBD7 | |
| Recombinant Human Kelch repeat and BTB domain-containing protein 7 protein (475-684AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction