missing translation for 'onlineSavingsMsg'
Learn More

KCC1/SLC12A4 Antibody, Novus Biologicals™

Goat Polyclonal Antibody

Brand:  Novus Biologicals NBP1-06043

Product Code. 18001854

  • € 528.00 / 0.10mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

KCC1/SLC12A4 Polyclonal specifically detects KCC1/SLC12A4 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

KCC1/SLC12A4
Polyclonal
Unconjugated
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Electroneutral potassium-chloride cotransporter 1, Erythroid K-Cl cotransporter 1, FLJ17069, FLJ40489, hKCC1, KCC1erythroid K:Cl cotransporter, K-Cl cotransporter, potassium/chloride cotransporter 1, solute carrier family 12 (potassium/chloride transporters), member 4, solute carrier family 12 member 4
Goat
Affinity Purified
RUO
6560
Human
IgG
Western Blot, Immunohistochemistry (Paraffin), ELISA
0.5 mg/mL
Western Blot 1:100-1:2000, Immunohistochemistry-Paraffin, Peptide ELISA Detection limit 1:4000
NP_005063.1
SLC12A4
Peptide with sequence C-DPSHAPDNFRE corresponding to internal region according to NP_005063.1.
0.1 mg
Primary
Cross reacts with Human. Expected from sequence similarity: Mouse, Rat, Dog.
Store at -20C. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only