missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ KCC3/SLC12A6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30600
This item is not returnable.
View return policy
Description
KCC3/SLC12A6 Polyclonal antibody specifically detects KCC3/SLC12A6 in Human samples. It is validated for Western Blot
Specifications
| KCC3/SLC12A6 | |
| Polyclonal | |
| Tris-citrate/phosphate buffer, pH 7 to 8 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| ACCPN, agenesis of corpus callosum and peripheral neuropathy (Andermann syndrome), DKFZp434D2135, EC 1.2.4.1, EC 6.1.1.3, Electroneutral potassium-chloride cotransporter 3, KCC3A, KCC3B, KCC3potassium chloride cotransporter 3, K-Cl cotransporter 3, potassium chloride cotransporter KCC3a-S3, potassium-chloride transporter-3a, potassium-chloride transporter-3b, solute carrier family 12 (potassium/chloride transporters), member 6, solute carrier family 12 member 6 | |
| Between 25 and 75 | |
| 100 μg | |
| Apoptosis, Cell Biology | |
| 9990 | |
| Store at 4°C. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction