Learn More
KCC4/SLC12A7 Antibody (891526), PE/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB9030PECY7
This item is not returnable.
View return policy
Description
KCC4/SLC12A7 Monoclonal antibody specifically detects KCC4/SLC12A7 in Human samples. It is validated for Flow Cytometry
Specifications
| KCC4/SLC12A7 | |
| Monoclonal | |
| PE-Cyanine7 | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 10723 | |
| Store at 4C in the dark. Do not freeze. | |
| IgG1 |
| Flow Cytometry | |
| 891526 | |
| Flow Cytometry | |
| DKFZp434F076, KCC4Electroneutral potassium-chloride cotransporter 4, K-Cl cotransporter 4, potassium/chloride transporter KCC4, solute carrier family 12 (potassium/chloride transporters), member 7, solute carrier family 12 member 7 | |
| NS0 mouse myeloma cell line transfected with human SLC12A7, Met1-Ser1083, Accession # Q9Y666 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |