missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KChIP2 Antibody (KCNIP2/7588) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-24038
This item is not returnable.
View return policy
Description
KChIP2 Monoclonal antibody specifically detects KChIP2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| KChIP2 | |
| Monoclonal | |
| 1 mg/mL | |
| Immunohistochemistry-Paraffin: 1-2 μg/mL | |
| A-type potassium channel modulatory protein 2, cardiac voltage gated potassium channel modulatory subunit, Cardiac voltage-gated potassium channel modulatory subunit, DKFZp566L1246, KChIP2, KCHIP2Kv channel-interacting protein 2, Kv channel interacting protein 2, MGC17241, potassium channel interacting protein 2, Potassium channel-interacting protein 2 | |
| Recombinant fragment (around aa1-270) of human KChIP2 protein (exact sequence is proprietary) | |
| 100 μg | |
| Vision | |
| 30819 | |
| Store at -20 to -80°C. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| KCNIP2/7588 | |
| Unconjugated | |
| 10 mM PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction