missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KChIP2 Antibody (KCNIP2/7589), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-24039AF532
This item is not returnable.
View return policy
Description
KChIP2 Monoclonal antibody specifically detects KChIP2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| KChIP2 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| A-type potassium channel modulatory protein 2, cardiac voltage gated potassium channel modulatory subunit, Cardiac voltage-gated potassium channel modulatory subunit, DKFZp566L1246, KChIP2, KCHIP2Kv channel-interacting protein 2, Kv channel interacting protein 2, MGC17241, potassium channel interacting protein 2, Potassium channel-interacting protein 2 | |
| Recombinant fragment (around aa1-270) of human KChIP2 protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Vision | |
| 30819 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| KCNIP2/7589 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction