missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KChIP2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-17026
This item is not returnable.
View return policy
Description
KChIP2 Polyclonal specifically detects KChIP2 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| KChIP2 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| A-type potassium channel modulatory protein 2, cardiac voltage gated potassium channel modulatory subunit, Cardiac voltage-gated potassium channel modulatory subunit, DKFZp566L1246, KChIP2, KCHIP2Kv channel-interacting protein 2, Kv channel interacting protein 2, MGC17241, potassium channel interacting protein 2, Potassium channel-interacting protein 2 | |
| Rabbit | |
| 31 kDa | |
| 0.1 mL | |
| Vision | |
| 30819 | |
| Human, Mouse | |
| IgG |
| Western Blot | |
| 1mg/mL | |
| Western Blot 1:500-1:3000 | |
| Q9NS61 | |
| KCNIP2 | |
| Recombinant protein encompassing a sequence within the center region of human KChIP2. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse. Expected to cross react based on sequence identity: Chicken (95%), Dog (99%), Monkey (99%), Rat (97%), Bovine (99%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction