missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ KCNH7 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15721651 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15721651 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15721651 Supplier Invitrogen™ Supplier No. PA553713

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: DNCKLRRRKL SFESEGEKEN STNDPEDSAD TIRHYQSSKR HFEEKKSRSS SFISSIDDEQ KPLFSGIVDS SPGIGKASGL DFEETVPTSG RMHIDKRSHS CKDITDMRSW ERENAHPQPE DSSPSALQRA A Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 73%.

Voltage-gated potassium (Kv) channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. This gene encodes a member of the potassium channel, voltage-gated, subfamily H. This member is a pore-forming (alpha) subunit. There are at least two alternatively spliced transcript variants derived from this gene and encoding distinct isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen KCNH7
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.6 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene KCNH7
Gene Accession No. Q9NS40
Gene Alias 9330137I11Rik; eag-related protein 3; erg3; ERG-3; ether-a-go-go-related gene potassium channel 3; ether-a-go-go-related protein 3; H Kv11.3; HERG3; KCNH7; Kv11.3; potassium channel erg3; potassium channel protein erg3; potassium channel subunit HERG-3; potassium channel, voltage gated eag related subfamily H, member 7; potassium voltage-gated channel subfamily H member 7; potassium voltage-gated channel, subfamily H (eag-related), member 7; voltage-gated potassium channel subunit Kv11.3
Gene Symbols KCNH7
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human KCNH7. Recombinant protein control fragment (Product #RP-91200).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 90134
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.