missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCNIP2 Monoclonal Antibody (OTI2G8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA807376S
This item is not returnable.
View return policy
Description
KCNIP2 Monoclonal antibody specifically detects KCNIP2 in Human samples. It is validated for Western BlotSpecifications
| KCNIP2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9NS61 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 30819 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI2G8 | |
| Unconjugated | |
| KCNIP2 | |
| A-type potassium channel modulatory protein 2; Cardiac voltage-gated potassium channel modulatory subunit; DKFZp566L1246; KCHIP2; KChlP2; Kcnip2; Kv channel interacting protein 2; Kv channel-interacting protein; Kv channel-interacting protein 2; MGC17241; potassium channel auxiliary subunit KCHIP2; potassium channel-interacting protein 2; potassium voltage-gated channel interacting protein 2; si:ch73-173h19.2 | |
| Full length human recombinant protein of human KCNIP2 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction