missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCNJ15, Mouse anti-Human, Clone: 1F8, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00003772-M13.100ug
This item is not returnable.
View return policy
Description
Sequence: MDAIHIGMSSTPLVKHTAGAGLKANRPRVMSKSGHSNVRIDKVDGIYLLYLQDLWTTVIDMKWRYKLTLFAATFVMTWFLFGVIYYAIAFIHGDLEPGEPISNHTPCIMKVDSLTGAFLFSLESQTTIGYGVRSITEECPHAIFLLVAQLVITTLIEIFITGTFLAKIARPKKRAETIKFSHCAVITKQNGKLCLVIQVANMRKSLLIQCQLSGKLLQTHVTKEGERILLNQATVKFHVDSSSESPFLILPMTFYHVLDETSPLRDLTPQNLKEKEFELVVLLNATVESTSAVCQSRTSYIPEEVYWGFEFVPVVSLSKNGKYVADFSQFEQIRKSPDCTFYCADSEKQQLEEKYRQEDQRERELRTLLLQQSNVSpecifications
| KCNJ15 | |
| Monoclonal | |
| Unconjugated | |
| potassium inwardly-rectifying channel, subfamily J, member 15 | |
| IRKK/KIR1.3/KIR4.2/MGC13584 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 1F8 | |
| In 1x PBS, pH 7.4 | |
| BC013327 | |
| KCNJ15 | |
| KCNJ15 (AAH13327, 1 a.a. ∼ 375 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 3772 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction