missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCNK12 Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81727IR
This item is not returnable.
View return policy
Description
KCNK12 Polyclonal antibody specifically detects KCNK12 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| KCNK12 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 755 | |
| potassium channel subfamily K member 12, potassium channel, subfamily K, member 12, tandem pore domain potassium channel THIK-2, THIK2, THIK-2, THIK-2THIK2Tandem pore domain halothane-inhibited potassium channel 2 | |
| KCNK12 antibody was raised against a 14 amino acid synthetic peptide near the carboxy terminus of human KCNK12. The immunogen is located within amino acids 320 - 370 of KCNK12. Amino Acid Squence: PGSRLRRRLAALGA | |
| 0.1 mL | |
| Signal Transduction | |
| 56660 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction