missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCNK13 Antibody, Alexa Fluor™ 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-41132AF488
This item is not returnable.
View return policy
Description
KCNK13 Polyclonal antibody specifically detects KCNK13 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| KCNK13 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 488 | |
| K2p13.1, potassium channel subfamily K member 13, potassium channel, subfamily K, member 13, tandem pore domain potassium channel THIK-1, THIK-1 | |
| Antibody was raised against a 15 amino acid synthetic peptide near the center of human KCNK13. The immunogen is located within amino acids 150 - 200 of KCNK13. Amino Acid Squence: RRGALPQESLKDAGQ | |
| 0.1 mL | |
| Signal Transduction | |
| 56659 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction