Learn More
Invitrogen™ KCNK9 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA588637
Description
Immunogen sequence: AGNRNSMVIH IPEEPRPSRP RYKADVPDLQ SVCSCTCYRS QDYGGRSVAP QNSFSAKLAP HYFHSISYKI EEISPSTLKN SLFPSPISSI SPGLHSFTDH QRLMKRRKSV; Positive Samples: SH-SY5Y, Mouse brain, Mouse intestine, Mouse pancreas; Cellular Location: Cell membrane, Multi-pass membrane protein.
KCNK9 is one of the members of the superfamily of potassium channel proteins containing two pore-forming P domains. This open channel is highly expressed in the cerebellum. It is inhibited by extracellular acidification and arachidonic acid, and strongly inhibited by phorbol 12-myristate 13-acetate.This gene encodes one of the members of the superfamily of potassium channel proteins containing two pore-forming P domains. This open channel is highly expressed in the cerebellum. It is inhibited by extracellular acidification and arachidonic acid, and strongly inhibited by phorbol 12-myristate 13-acetate.
Specifications
| KCNK9 | |
| Polyclonal | |
| Unconjugated | |
| Kcnk9 | |
| acid-sensitive potassium channel protein TASK-3; K2p9.1; Kcnk9; KT3.2; MGC138268; MGC138270; Potassium channel subfamily K member 9; potassium channel subfamily K member 9 (Task-3); potassium channel TASK3; potassium channel, subfamily K, member 9; potassium channel, subfamily K, member 9 (Task-3); potassium channel, two pore domain subfamily K, member 9; potassium two pore domain channel subfamily K member 9; TASK3; TASK-3; TWIK-related acid-sensitive K(+) channel 3; two pore K(+) channel KT3.2; Two pore potassium channel KT3.2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 223604, 51305 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.62 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q3LS21, Q9NPC2 | |
| Kcnk9 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 265-374 of human KCNK9 (NP_0012694631). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.