missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KCNK9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114350
This item is not returnable.
View return policy
Description
KCNK9 Polyclonal Antibody for Western Blot, IHC (P)
KCNK9 is one of the members of the superfamily of potassium channel proteins containing two pore-forming P domains. This open channel is highly expressed in the cerebellum. It is inhibited by extracellular acidification and arachidonic acid, and strongly inhibited by phorbol 12-myristate 13-acetate.This gene encodes one of the members of the superfamily of potassium channel proteins containing two pore-forming P domains. This open channel is highly expressed in the cerebellum. It is inhibited by extracellular acidification and arachidonic acid, and strongly inhibited by phorbol 12-myristate 13-acetate.
Specifications
| KCNK9 | |
| Polyclonal | |
| Unconjugated | |
| Kcnk9 | |
| acid-sensitive potassium channel protein TASK-3; K2p9.1; Kcnk9; KT3.2; MGC138268; MGC138270; Potassium channel subfamily K member 9; potassium channel subfamily K member 9 (Task-3); potassium channel TASK3; potassium channel, subfamily K, member 9; potassium channel, subfamily K, member 9 (Task-3); potassium channel, two pore domain subfamily K, member 9; potassium two pore domain channel subfamily K member 9; TASK3; TASK-3; TWIK-related acid-sensitive K(+) channel 3; two pore K(+) channel KT3.2; Two pore potassium channel KT3.2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51305, 84429 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q9ES08, Q9NPC2 | |
| Kcnk9 | |
| Synthetic peptide within rat KCNK9 aa 41-90. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction