missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KCNMB2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA577262
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
MaxiK channels are large conductance, voltage and calcium-sensitive potassium channels which are fundamental to the control of smooth muscle tone and neuronal excitability. MaxiK channels can be formed by 2 subunits: the pore-forming alpha subunit and the modulatory beta subunit. The protein encoded by this gene is an auxiliary beta subunit which decreases the activation time of MaxiK alpha subunit currents. Alternative splicing results in multiple transcript variants of this gene. Additional variants are discussed in the literature, but their full length nature has not been described.
Specifications
| KCNMB2 | |
| Polyclonal | |
| Unconjugated | |
| Kcnmb2 | |
| 2700049B16Rik; 3110031N04Rik; big potassium channel beta subunit 2; BK channel beta subunit 2; BK channel subunit beta-2; BKbeta2; calcium-activated potassium channel beta 2; calcium-activated potassium channel subunit beta-2; Calcium-activated potassium channel, subfamily M subunit beta-2; charybdotoxin receptor subunit beta-2; hbeta2; Hbeta3; hCG1646471; k(VCA)beta-2; Kcmb2; Kcnmb2; large conductance calcium-activated potassium channel beta 2 subunit; large-conductance Ca2+-activated K+ channel beta2 subunit; maxi K channel subunit beta-2; MaxiK channel beta 2 subunit; MaxiK channel beta-subunit 2; potassium calcium-activated channel subfamily M regulatory beta subunit 2; potassium channel subfamily M regulatory beta subunit 2; potassium large conductance calcium-activated channel, subfamily M, beta member 2; rbeta3; Slo-beta2; Slo-beta-2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10242, 294961, 72413 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q811Q0, Q9CZM9, Q9Y691 | |
| Kcnmb2 | |
| Synthetic peptide corresponding to amino acids 154-203 of Human KCNMB2. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction