Learn More
Invitrogen™ KCNMB2 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5106621
Product Code. 16683055
  • € 491.00 / 100µL
Description
Antibody detects endogenous levels of total KCNMB2.
MaxiK channels are large conductance, voltage and calcium-sensitive potassium channels which are fundamental to the control of smooth muscle tone and neuronal excitability. MaxiK channels can be formed by 2 subunits: the pore-forming alpha subunit and the modulatory beta subunit. The protein encoded by this gene is an auxiliary beta subunit which decreases the activation time of MaxiK alpha subunit currents. Alternative splicing results in multiple transcript variants of this gene. Additional variants are discussed in the literature, but their full length nature has not been described.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
KCNMB2
Polyclonal
Unconjugated
Kcnmb2
2700049B16Rik; 3110031N04Rik; big potassium channel beta subunit 2; BK channel beta subunit 2; BK channel subunit beta-2; BKbeta2; calcium-activated potassium channel beta 2; calcium-activated potassium channel subunit beta-2; Calcium-activated potassium channel, subfamily M subunit beta-2; charybdotoxin receptor subunit beta-2; hbeta2; Hbeta3; hCG1646471; k(VCA)beta-2; Kcmb2; Kcnmb2; large conductance calcium-activated potassium channel beta 2 subunit; large-conductance Ca2+-activated K+ channel beta2 subunit; maxi K channel subunit beta-2; MaxiK channel beta 2 subunit; MaxiK channel beta-subunit 2; potassium calcium-activated channel subfamily M regulatory beta subunit 2; potassium channel subfamily M regulatory beta subunit 2; potassium large conductance calcium-activated channel, subfamily M, beta member 2; rbeta3; Slo-beta2; Slo-beta-2
Rabbit
Affinity chromatography
RUO
10242, 294961, 72413
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q811Q0, Q9CZM9, Q9Y691
Kcnmb2
A synthesized peptide derived from human KCNMB2(Accession Q9Y691), corresponding to amino acid residues S155-C205.
100 μL
Primary
Human, Mouse, Rat