missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCNQ4, Mouse anti-Human, Clone: 2H6, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00009132-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: AREKGDKGPSDAEVVDEISMMGRVVKVEKQVQSIEHKLDLLLGFYSRCLRSGTSASLGAVQVPLFDPDITSDYHSPVDHEDISVSAQTLSISRSVSTNMDSpecifications
| KCNQ4 | |
| Monoclonal | |
| Unconjugated | |
| potassium voltage-gated channel, KQT-like subfamily, member 4 | |
| DFNA2/KV7.4 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody |
| ELISA, Western Blot | |
| 2H6 | |
| In 1x PBS, pH 7.4 | |
| NM_004700 | |
| KCNQ4 | |
| KCNQ4 (NP_004691, 596 a.a. ∼ 695 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 9132 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG3 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction