Learn More
KCNT2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-14146
 View more versions of this product
Product Code. 18761734
  • € 589.00 / 0.10mL
Description
KCNT2 Polyclonal specifically detects KCNT2 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polyclonal
Rabbit
0.1 mL
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.