missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KCNV1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5144946
This item is not returnable.
View return policy
Description
Immunogen sequence: LCALSFLQEI QYWGIDELSI DSCCRDRYFR RKELSETLDF KKDTEDQESQ HESEQDFSQG PCPTVRQKLW NILEK Antigen sequence identity: Mouse 96%; Rat 97%.
Voltage-gated potassium (Kv) channels represent the most complex class of voltage-gated ion channels from both functional and structural standpoints. Their diverse functions include regulating neurotransmitter release, heart rate, insulin secretion, neuronal excitability, epithelial electrolyte transport, smooth muscle contraction, and cell volume. This gene encodes a member of the potassium voltage-gated channel subfamily V. This protein is essentially present in the brain, and its role might be to inhibit the function of a particular class of outward rectifier potassium channel types.
Specifications
| KCNV1 | |
| Polyclonal | |
| Unconjugated | |
| KCNV1 | |
| 2700023A03Rik; HNKA; KCNB3; KCNV1; KV2.3; kv2.3r; Kv8.1; neuronal potassium channel alpha subunit; neuronal potassium channel alpha subunit HNKA; potassium channel Kv8.1; potassium channel Kv8.1 homolog; potassium channel, subfamily V, member 1; potassium channel, voltage gated modifier subfamily V, member 1; potassium channel, voltage-gated modifier subfamily V, member 1; potassium voltage-gated channel modifier subfamily V member 1; potassium voltage-gated channel subfamily V member 1; vibe; Voltage-gated potassium channel subunit Kv2.3r; voltage-gated potassium channel subunit Kv8.1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 27012 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid | |
| IgG |
| Immunocytochemistry | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6PIU1 | |
| KCNV1 | |
| Recombinant Protein Epitope Signature Tag (PrEST) antigen sequence. Antigen range: 128-202. Recombinant protein control fragment (Product #RP-110121). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction