missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCP Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-81728CL1
This item is not returnable.
View return policy
Description
KCP Polyclonal antibody specifically detects KCP in Human,Mouse,Rat samples. It is validated for ELISA,Western Blot
Specifications
| KCP | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 375616 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| ELISA, Western Blot | |
| CoraFluor 1 | |
| CRIM2, KCP1, kielin/chordin-like protein, NET67 | |
| KCP antibody was raised against a 15 amino acid synthetic peptide near the carboxy terminus of human KCP. The immunogen is located within amino acids 1320 - 1370 of KCP. Amino Acid Squence: RAAGYRARREANAR | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction