missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCTD1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-21290-100ul
This item is not returnable.
View return policy
Description
KCTD1 Polyclonal antibody specifically detects KCTD1 in Human samples. It is validated for Western Blot, Immunofluorescence
Specifications
| KCTD1 | |
| Polyclonal | |
| Western Blot 0.04-0.4 ug/ml, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml | |
| BTB/POZ domain-containing protein KCTD1, C18orf5, potassium channel tetramerisation domain containing 1 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: SKSSLISIRSSLNRYLNEPPYCRTLDLTKDPELRSANLTLAAVIRKLEEQGAGPVVQKQAITRADLRKLYTSSVFSTNTPFGLLNKVWFETCMY | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS, pH 7.2, 40% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 284252 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction