missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCTD12 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-80568B
This item is not returnable.
View return policy
Description
KCTD12 Polyclonal antibody specifically detects KCTD12 in Human samples. It is validated for Western Blot
Specifications
| KCTD12 | |
| Polyclonal | |
| Western Blot | |
| BTB/POZ domain-containing protein KCTD12, C13orf2, chromosome 13 open reading frame 2, FLJ33073, KIAA1778PFET1PFETIN, Pfetin, potassium channel tetramerisation domain containing 12, Predominantly fetal expressed T1 domain | |
| The immunogen this antibody was made to, maps to a region between residue 125 and 175 of human Predominantly Fetal Expressed T1 Domain using the numbering given in entry NP_612453.1 (GeneID 115207). | |
| 0.1 mL | |
| Cancer, Neuroscience | |
| 115207 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| Biotin | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction