missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCTD13 Monoclonal Antibody (OTI1A4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
€ 242.00 - € 540.87
Specifications
| Antigen | KCTD13 |
|---|---|
| Clone | OTI1A4 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17530434
|
Origene Technologies
TA808802S |
30 μL |
€ 242.00
30µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17510434
|
Origene Technologies
TA808802 |
100 μL |
€ 540.87
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KCTD13 Monoclonal antibody specifically detects KCTD13 in Human samples. It is validated for Western BlotSpecifications
| KCTD13 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| Q8WZ19 | |
| IgG1 | |
| -20° C, Avoid Freeze/Thaw Cycles |
| OTI1A4 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| 253980 | |
| Primary | |
| KCTD13 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title