missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCTD14 Monoclonal Antibody (OTI2A6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502655
This item is not returnable.
View return policy
Description
KCTD14 is a protein coding gene.Specifications
| KCTD14 | |
| Monoclonal | |
| 0.89 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9BQ13 | |
| KCTD14 | |
| Full length human recombit protein of human KCTD14 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI2A6 | |
| Unconjugated | |
| KCTD14 | |
| potassium channel tetramerization domain containing 14 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 65987 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction