Learn More
KCTD14 Mouse anti-Human, Clone: OTI1D11, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF502534
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
KCTD14 is a protein coding gene.Specifications
| KCTD14 | |
| Monoclonal | |
| Unconjugated | |
| KCTD14 | |
| potassium channel tetramerization domain containing 14 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 65987 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI1D11 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q9BQ13 | |
| KCTD14 | |
| Full length human recombit protein of human KCTD14 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.