Learn More
Invitrogen™ KCTD16 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114105
Description
Immunogen sequence: KREAEYFQLP DLVKLLTPDE IKQSPDEFCH SDFEDASQGS DTRICPPSSL For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 93%; Guinea Pig: 79%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
The BTB (Broad-Complex, Tramtrack and Bric a brac) domain, also known as the POZ (Poxvirus and Zinc finger) domain, is an N-terminal homodimerization domain that contains multiple copies of kelch repeats and/or C2H2-type zinc fingers. Proteins that contain BTB domains are thought to be involved in transcriptional regulation via control of chromatin structure and function. KCTD16 (potassium channel tetramerisation domain containing 16), also known as BTB/POZ domain-containing protein KCTD16, is a 428 amino acid protein that contains one BTB (POZ) domain. An auxiliary subunit of GABAB R1 and GABAB R2, KCTD16 increases agonist potency and alters the G-protein signaling of the receptors by accelerating onset and promoting desensitization.
Specifications
| KCTD16 | |
| Polyclonal | |
| Unconjugated | |
| KCTD16 | |
| 4930434H12Rik; BTB/POZ domain-containing protein KCTD16; Gm1267; KCTD16; KIAA1317; potassium channel tetramerisation domain containing 16; potassium channel tetramerization domain containing 16; potassium channel tetramerization domain-containing protein 16 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 57528 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q68DU8 | |
| KCTD16 | |
| Synthetic peptide directed towards the N terminal region of human KCTD16 (aa 107-156). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.