missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KCTD5 Antibody (OTI3C8), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71955F
This item is not returnable.
View return policy
Description
KCTD5 Monoclonal specifically detects KCTD5 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Immunohistochemistry.
Specifications
| KCTD5 | |
| Monoclonal | |
| FITC | |
| PBS with 0.05% Sodium Azide | |
| KCTD5 | |
| Full length recombinant protein of human KCTD5 (NP_061865) produced in HEK293T cells. | |
| 0.1 mL | |
| 54442.0 | |
| Store at 4°C in the dark. |
| Immunohistochemistry | |
| OTI3C8 | |
| Immunohistochemistry | |
| BTB/POZ domain-containing protein KCTD5, FLJ20040, potassium channel tetramerisation domain containing 5 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction