Learn More
KCTD9 Mouse anti-Human, Clone: OTI2G8, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF807664
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Substrate-specific adapter of a BCR (BTB-CUL3-RBX1) E3 ubiquitin-protein ligase complex, which mediates the ubiquitination of target proteins, leading to their degradation by the proteasome.Specifications
| KCTD9 | |
| Monoclonal | |
| Unconjugated | |
| KCTD9 | |
| BTBD27 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 54793 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2G8 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q7L273 | |
| KCTD9 | |
| Human recombit protein fragment corresponding to amino acids 1-223 of human KCTD9 produced in E.coli. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.