missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ KCTD9 Recombinant Rabbit Monoclonal Antibody (23GB6305), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
€ 195.00 - € 468.00
Specifications
| Antigen | KCTD9 |
|---|---|
| Clone | 23GB6305 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088139
![]() |
Invitrogen™
MA553016 |
100 μL |
€ 468.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30087787
|
Invitrogen™
MA553017 |
20 μL |
€ 195.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
KCTD9 Recombinant Monoclonal antibody specifically detects KCTD9 in Human samples. It is validated for Flow Cytometry, Western BlotSpecifications
| KCTD9 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q7L273 | |
| 54793 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB6305 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| BTB/POZ domain-containing protein KCTD9; BTBD27; KCTD9; potassium channel tetramerisation domain containing 9; potassium channel tetramerization domain containing 9 | |
| A synthesized peptide derived from human KCTD9 (61-85AA). | |
| Primary | |
| -20°C | |
| Kctd9 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title