Learn More
KD Scientific Stainless-Steel Syringes
Product Code. 15114184
Click to view available options
Volume (Metric):
2.5 mL
8 mL
20 mL
50 mL
100 mL
200 mL
Product Type:
Luer-Lok Syringe
Swagelok 1/16 in. Syringe
Swagelok 1/4 in. Syringe
Swagelok 1/8 in. Syringe
Unit Size:
Each
Product Code. 15114184
Brand: KD Scientific 780814
Only null left
Add to basket
Add to basket
Durable, electron-beam construction withstands high force, high pressure applications
Eliminates hazards of glass syringe breakage.
  • Compatible with most syringe pumps
  • Fully autoclavable for reuse
  • Adaptable to Luer Lock or Swagelok™ fittings
  • Resistant to most chemicals
Specifications
Autoclavable Autoclavable
Material Stainless Steel
Diameter (English) Inner 1-37/100 in.
Diameter (English) Outer 1-1/5 in.
Diameter (Metric) Inner 34.7 mm
Diameter (Metric) Outer 38.1 mm
Product Type Swagelok 1/4 in. Syringe
Max. Pressure 1500 psi
Volume (Metric) 100 mL
For Use With (Equipment) Syringe Pump