Learn More
KD Scientific Stainless-Steel Syringes
Durable, electron-beam construction withstands high force, high pressure applications
Brand: KD Scientific 780808
Additional Details : Weight : 0.17000kg
Description
Eliminates hazards of glass syringe breakage.
- Compatible with most syringe pumps
- Fully autoclavable for reuse
- Adaptable to Luer Lock or Swagelok™ fittings
- Resistant to most chemicals
Specifications
20 mL | |
22/25 in. | |
19 mm | |
Stainless Steel (316) | |
1500 psi |
3/4 in. | |
22.3 mm | |
Syringe Pump | |
Swagelok 1/8 in. Syringe |