Learn More
KD Scientific Stainless-Steel Syringes
missing translation for 'orderingAttributeHoverText'
Volume (Metric):
2.5 mL
8 mL
20 mL
50 mL
100 mL
200 mL
Product Type:
Luer-Lok Syringe
Swagelok 1/16 in. Syringe
Swagelok 1/4 in. Syringe
Swagelok 1/8 in. Syringe
missing translation for 'unitSize'
Each
Description
Eliminates hazards of glass syringe breakage.
- Compatible with most syringe pumps
- Fully autoclavable for reuse
- Adaptable to Luer Lock or Swagelok™ fittings
- Resistant to most chemicals
Specifications
Specifications
| Autoclavable | Autoclavable |
| Material | Stainless Steel |
| Diameter (English) Inner | 19/100 in. |
| Diameter (English) Outer | 1/2 in. |
| Diameter (Metric) Inner | 4.8 mm |
| Diameter (Metric) Outer | 12.7 mm |
| Product Type | Swagelok 1/16 in. Syringe |
| Max. Pressure | 7000 psi |
| Volume (Metric) | 2.5 mL |
| For Use With (Equipment) | Syringe Pump |