missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KDELR1 Antibody (KR-10), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12873AF405
This item is not returnable.
View return policy
Description
KDELR1 Monoclonal specifically detects KDELR1 in Human, Mouse, Rat, Porcine, Bovine, Canine, Chicken, Drosophila, Hamster, Mammal, Monkey, Rabbit, Sheep, Xenopus samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| KDELR1 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| ER lumen protein retaining receptor 1, ERD2, ERD2.1KDEL receptor 1, HDEL, KDEL (Lys-Asp-Glu-Leu) endoplasmic reticulum protein retention receptor 1, KDEL endoplasmic reticulum protein retention receptor 1, PM23, Putative MAPK-activating protein PM23 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 10945 | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Chicken, Drosophila, Hamster, Mammalia, Monkey, Rabbit, Sheep, Xenopus species | |
| Purified |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| KR-10 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| KDELR1 | |
| A 21 residue synthetic peptide (amino acids 192-212) based on the bovine KDEL receptor and the peptide coupled to KLH | |
| 0.1 mL | |
| Primary | |
| Detects approx 25kDa. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction