missing translation for 'onlineSavingsMsg'
Learn More

KDM2B Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP3-35627-20ul

 View more versions of this product

Product Code. 30232127

  • € 190.00 / 20µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

KDM2B Polyclonal antibody specifically detects KDM2B in Human samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

KDM2B
Polyclonal
Western Blot 1:500 - 1:2000, ELISA
[Histone-H3]-lysine-36 demethylase 1B, CXXC2F-box protein FBL10, CXXC-type zinc finger protein 2, Fbl10, F-box and leucine-rich repeat protein 10JEMMA (Jumonji domain, EMSY-interactor, methyltransferase motif) protein, FBXL10, JHDM1BF-box/LRR-repeat protein 10, JmjC domain-containing histone demethylation protein 1B, jumonji C domain-containing histone demethylase 1B, Jumonji domain-containing EMSY-interactor methyltransferase motif protein, lysine (K)-specific demethylase 2B, lysine-specific demethylase 2B, PCCX2EC 1.14.11.27, protein containing CXXC domain 2, Protein JEMMA, Protein-containing CXXC domain 2
A synthetic peptide corresponding to a sequence within amino acids 600-700 of human KDM2B (XP_011537177.1).,, Sequence:, SGCSWIAVSALCSSSCPLLRTLDVQWVEGLKDAQMRDLLSPPTDNRPGQMDNRSKLRNIVELRLAGLDITDASLRLIIRHMPLLSKLHLSYCNHVTDQSIN
20 μL
Primary
Human
Purified
ELISA, Western Blot
Unconjugated
PBS (pH 7.3), 50% glycerol
Rabbit
Affinity purified
RUO
84678
Store at -20°C. Avoid freeze-thaw cycles.
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.