missing translation for 'onlineSavingsMsg'
Learn More

KDM4D-IN-1, MedChemExpress

Product Code. p-200100295
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16450037

Brand: Medchem Express HY101928

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress KDM4D-IN-1 is a new histone lysine demethylase 4D (KDM4D) inhibitor with an IC50 value of 0.41±0.03 μM.

KDM4D-IN-1 is a new histone lysine demethylase 4D (KDM4D) inhibitor with an IC50 value of 0.41±0.03 μM.

Specifications

Chemical Name or Material KDM4D-IN-1
CAS 2098902-68-0
Color Earth Yellow
Purity Grade Notes Research
Molecular Formula C11H7N5O
Quantity 50 mg
Solubility Information DMSO : 7.14 mg/mL (31.70 mM; Need ultrasonic)
SMILES OC1=NC2=C(C#N)C(C)=NN2C3=C1C=CC=N3
Molecular Weight (g/mol) 225.21
Formula Weight 225.21
Percent Purity 99.91%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.