Learn More
KDM4D Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 22591-1-AP-150UL
This item is not returnable.
View return policy
Description
JMJD2D belongs to JMJD2 family that is consist of four cancer-associated genes.Specifications
| KDM4D | |
| Polyclonal | |
| Unconjugated | |
| KDM4D | |
| JHDM3D, JMJD2D, KDM4D, lysine (K) demethylase 4D, Lysine specific demethylase 4D | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 55693 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.4 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q6B0I6 | |
| KDM4D | |
| KDM4D Fusion Protein Ag18436 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |