missing translation for 'onlineSavingsMsg'
Learn More

KDM5A-IN-1, MedChemExpress

Product Code. 16482738
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16482738

Brand: Medchem Express HY100014

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress KDM5A-IN-1 is a potent, orally bioavailable pan-histone lysine demethylases 5 (KDM5) inhibitor with IC50s of 45 nM, 56 nM and 55 nM for KDM5A, KDM5B and KDM5C, respectively, and with an EC50 value of 960 nM for PC9 H3K4Me3. KDM5A-IN-1 is significantly less potent against other KDM5B enzymes (1A, 2B, 3B, 4C, 6A, 7B).

KDM5A-IN-1 is a potent, orally bioavailable pan-histone lysine demethylases 5 (KDM5) inhibitor with IC50s of 45 nM, 56 nM and 55 nM for KDM5A, KDM5B and KDM5C, respectively, and with an EC50 value of 960 nM for PC9 H3K4Me3. KDM5A-IN-1 is significantly less potent against other KDM5B enzymes (1A, 2B, 3B, 4C, 6A, 7B).

Specifications

Chemical Name or Material KDM5A-IN-1
Physical Form Powder
CAS 1905481-36-8
Color Off-White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C15H22N4O2
For Use With (Application) Cancer-programmed cell death
Solubility Information DMSO : ≥ 50 mg/mL (172.20 mM)
SMILES O=C(C1CC1)N[C@@H]2CCN(C(C3=CC(C(C)C)=NN3)=O)C2
Molecular Weight (g/mol) 290.36
Formula Weight 290.36
Percent Purity 95.71%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.