missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KDM5A-IN-1, MedChemExpress
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
Description
KDM5A-IN-1 is a potent, orally bioavailable pan-histone lysine demethylases 5 (KDM5) inhibitor with IC50s of 45 nM, 56 nM and 55 nM for KDM5A, KDM5B and KDM5C, respectively, and with an EC50 value of 960 nM for PC9 H3K4Me3. KDM5A-IN-1 is significantly less potent against other KDM5B enzymes (1A, 2B, 3B, 4C, 6A, 7B).
Specifications
Specifications
| Chemical Name or Material | KDM5A-IN-1 |
| Physical Form | Powder |
| CAS | 1905481-36-8 |
| Color | Off-White |
| Quantity | 1 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C15H22N4O2 |
| For Use With (Application) | Cancer-programmed cell death |
| Solubility Information | DMSO : ≥ 50 mg/mL (172.20 mM) |
| SMILES | O=C(C1CC1)N[C@@H]2CCN(C(C3=CC(C(C)C)=NN3)=O)C2 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction