missing translation for 'onlineSavingsMsg'
Learn More
Learn More
KDM6A Mouse anti-Human, Rat, Clone:6E2D6, Abnova™
Mouse monoclonal antibody raised against recombinant human KDM6A.
Brand: Abnova MAB17186.100ug
This item is not returnable.
View return policy
Specifications
| KDM6A | |
| Monoclonal | |
| Unconjugated | |
| ELISA (1:10000) Western Blot (1:500-1:2000) Immunohistochemistry Immunocytochemistry Flow Cytometry The optimal working dilution should be determined by the end user. |
|
| KDM6A | |
| Mouse | |
| 100 μg | |
| Human, Rat | |
| Antibody | |
| IgG2a |
| ELISA, Western Blot | |
| 6E2D6 | |
| lysine (K)-specific demethylase 6A | |
| In PBS (0.05% sodium azide) | |
| UTX, KABUK2, bA386N14.2 | |
| Recombinant protein corresponding to amino acid 1252-1401 of human KDM6A from E. coli. | |
| 7403 | |
| Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction